Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF426 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318055.1 | NP_001304984.1 | 4 | 371 | ||
NM_001318056.1 | NP_001304985.1 | 4 | 371 | |||
NM_024106.2 | NP_077011.1 | 4 | 371 | |||
GenBank mRNA | AK095759.1 | - | 2 | 426 | ||
BC001791.1 | - | 4 | 353 | 3543910 | ||
BE315219.1 | - | 4 | 316 | |||
BP245135.1 | - | 4 | 371 |