Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRAPPC6A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270891.1 | NP_001257820.1 | 6 | 491 | ||
NM_001270892.1 | NP_001257821.1 | 5 | 465 | |||
NM_001270893.1 | NP_001257822.1 | 5 | 423 | |||
NM_024108.2 | NP_077013.1 | 6 | 533 | |||
XM_017027295.1 | XP_016882784.1 | 6 | 629 | |||
GenBank mRNA | AF161407.1 | - | 6 | 485 | ||
BC001907.1 | - | 6 | 517 | 3534111 | ||
BC004450.1 | - | 6 | 518 | 2820830 | ||
BI462720.1 | - | 5 | 420 | |||
CN256686.1 | - | 5 | 460 | |||
CN256687.1 | - | 6 | 503 | |||
FJ418644.1 | - | 6 | 473 |