Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C11orf49 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001003676.2 | NP_001003676.1 | 7 | 810 | ||
NM_001003677.2 | NP_001003677.1 | 7 | 810 | |||
NM_001003678.2 | NP_001003678.1 | 7 | 810 | |||
NM_001278222.1 | NP_001265151.1 | 6 | 707 | |||
NM_024113.4 | NP_077018.1 | 7 | 810 | |||
NR_103471.1 | 8 | 1055 | ||||
NR_103472.1 | 6 | 679 | ||||
XM_006718315.1 | XP_006718378.1 | 7 | 801 | |||
XM_011520364.2 | XP_011518666.1 | 6 | 872 | |||
XM_011520365.2 | XP_011518667.1 | 7 | 801 | |||
XM_017018276.1 | XP_016873765.1 | 7 | 801 | |||
XM_017018277.1 | XP_016873766.1 | 6 | 620 | |||
XM_017018278.1 | XP_016873767.1 | 8 | 984 | |||
XM_017018279.1 | XP_016873768.1 | 8 | 1047 | |||
XM_017018280.1 | XP_016873769.1 | 9 | 1708 | |||
XM_017018281.1 | XP_016873770.1 | 8 | 1677 | |||
XM_017018282.1 | XP_016873771.1 | 8 | 984 | |||
GenBank mRNA | AK025863.1 | - | 7 | 719 | ||
AK300813.1 | - | 6 | 617 | |||
AK316318.1 | - | 8 | 1047 | |||
AL136575.1 | - | 7 | 729 | |||
BC001860.2 | - | 7 | 756 | 3534541 | ||
BC009455.1 | - | 7 | 731 | 3535652 | ||
BC013988.2 | - | 7 | 721 | 3349493 | ||
BC052291.1 | - | 7 | 723 | 5936112 | ||
CK724853.1 | - | 3 | 429 | |||
CR457351.1 | - | 7 | 693 |