Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAPKAP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001006617.1 | NP_001006618.1 | 7 | 1230 | ||
NM_001006618.1 | NP_001006619.1 | 7 | 1230 | |||
NM_001006619.1 | NP_001006620.1 | 7 | 1230 | |||
NM_001006620.1 | NP_001006621.1 | 6 | 902 | |||
NM_001006621.1 | NP_001006622.1 | 5 | 812 | |||
NM_024117.3 | NP_077022.1 | 7 | 1230 | |||
XM_011519005.1 | XP_011517307.1 | 8 | 1371 | |||
XM_011519006.2 | XP_011517308.1 | 8 | 1371 | |||
XM_011519007.1 | XP_011517309.1 | 8 | 1371 | |||
XM_011519008.1 | XP_011517310.1 | 8 | 1371 | |||
XM_011519009.1 | XP_011517311.1 | 8 | 1371 | |||
XM_011519010.2 | XP_011517312.1 | 4 | 548 | |||
XM_017015126.1 | XP_016870615.1 | 7 | 1119 | |||
XM_017015127.1 | XP_016870616.1 | 7 | 1119 | |||
GenBank mRNA | AK290060.1 | - | 7 | 1043 | ||
AK295364.1 | - | 4 | 536 | |||
AK298841.1 | - | 3 | 304 | |||
AK309507.1 | - | 4 | 497 | |||
AK310361.1 | - | 4 | 536 | |||
AK316149.1 | - | 4 | 475 | |||
AL833042.1 | - | 6 | 863 | |||
AY524429.1 | - | 7 | 1227 | |||
AY524430.1 | - | 7 | 1227 | |||
AY524431.1 | - | 7 | 1227 | |||
AY524432.1 | - | 7 | 1227 | |||
AY633624.1 | - | 7 | 1197 | |||
AY633625.1 | - | 7 | 1204 | |||
AY633626.1 | - | 7 | 1204 | |||
BC002326.2 | - | 7 | 1199 | 2823015 | ||
BC003044.1 | - | 7 | 1199 | 2823015 | ||
M37191.1 | - | 6 | 675 |