Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DHX58 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024119.2 | NP_077024.2 | 12, 13 | 1977 | ||
XM_017025059.1 | XP_016880548.1 | 11, 12 | 1910 | |||
XM_017025060.1 | XP_016880549.1 | 12, 13 | 1973 | |||
GenBank mRNA | AK021416.1 | - | 12, 13 | 1977 | ||
BC014949.1 | - | 12, 13 | 1914 | 4865798 |