Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MBOAT7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001146056.2 | NP_001139528.1 | 6 | 1434 | ||
NM_001146083.2 | NP_001139555.1 | 7 | 1513 | |||
NM_024298.4 | NP_077274.3 | 8 | 1640 | |||
XM_011527299.2 | XP_011525601.1 | 8 | 1844 | |||
XM_011527300.2 | XP_011525602.1 | 8 | 1235 | |||
GenBank mRNA | AF211969.1 | - | 2 | 94 | ||
AK055908.1 | - | 8 | 1573 | |||
AK122789.1 | - | 9 | 1358 | |||
AK123290.1 | - | 2 | 2087 | |||
AK293199.1 | - | 6 | 1044 | |||
AK298689.1 | - | 6 | 1120 | |||
AK311493.1 | - | 7 | 916 | |||
BC002512.2 | - | 7 | 1090 | |||
BC003164.1 | - | 8 | 1171 | 2958347 | ||
BC015857.1 | - | 7 | 1077 | 4671596 | ||
EU016381.1 | - | 7 | 1092 | |||
S82470.1 | - | 6 | 946 |