Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHCHD7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011667.2 | NP_001011667.1 | 4, 5 | 561 | ||
NM_001011668.2 | NP_001011668.1 | 4, 5 | 517 | |||
NM_001011669.2 | NP_001011669.1 | 4, 5 | 541 | |||
NM_001011670.2 | NP_001011670.1 | 3, 4 | 413 | |||
NM_001011671.2 | NP_001011671.1 | 3, 4 | 369 | |||
NM_024300.4 | NP_077276.2 | 4, 5 | 497 | |||
NR_133937.1 | 3, 4 | 409 | ||||
GenBank mRNA | AK095922.1 | - | 3, 4 | 218 | ||
AK098285.1 | - | 2, 3 | 3020 | |||
AK225982.1 | - | 4, 5 | 356 | |||
AK290088.1 | - | 4, 5 | 346 | |||
AY070434.1 | - | 4, 5 | 389 | |||
BC002546.2 | - | 4, 5 | 336 | 3139026 | ||
BM695222.1 | - | 3, 4 | 253 | |||
BX473986.1 | - | 4, 5 | 399 |