Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RBM42 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001319113.1 | NP_001306042.1 | 9 | 1412 | ||
NM_024321.4 | NP_077297.2 | 10 | 1499 | |||
XM_005259257.2 | XP_005259314.1 | 10 | 1473 | |||
GenBank mRNA | BC002868.2 | - | 9 | 1351 | 3941786 | |
BC004204.2 | - | 10 | 1437 | 3532802 | ||
BC031682.1 | - | 9 | 1376 | 5163682 | ||
BU732481.1 | - | 1 | 229 |