Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IRX6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024335.2 | NP_077311.2 | 2 | 636 | ||
XM_005256137.2 | XP_005256194.1 | 2 | 1233 | |||
GenBank mRNA | AK125053.1 | - | 2 | 1435 | ||
AK309149.1 | - | 2 | 269 | |||
AY335937.1 | - | 2 | 636 | |||
BC136573.1 | - | 2 | 377 | 9020563 | ||
BC136576.1 | - | 2 | 377 | 9020566 |