Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZXDC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040653.3 | NP_001035743.1 | 6 | 1867 | ||
NM_025112.4 | NP_079388.3 | 6 | 1867 | |||
NR_104249.1 | 6 | 1867 | ||||
XM_005247757.3 | XP_005247814.1 | 6 | 1878 | |||
XM_006713741.2 | XP_006713804.1 | 6 | 1879 | |||
XM_011513119.2 | XP_011511421.1 | 5 | 1715 | |||
GenBank mRNA | AK023923.1 | - | 3 | 569 | ||
AK092101.1 | - | 6 | 1228 | |||
AY277596.1 | - | 2 | 429 | |||
AY936556.1 | - | 6 | 1827 | |||
BC002940.2 | - | 5 | 805 | |||
BC012729.2 | - | 5 | 805 | |||
BC063713.1 | - | 6 | 935 | |||
BC073859.1 | - | 4 | 729 | |||
DQ079590.1 | - | 6 | 1867 | |||
FJ980275.1 | - | 6 | 1080 |