Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BCL2L14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030766.1 | NP_110393.1 | 2 | 216 | ||
NM_138722.1 | NP_620048.1 | 2 | 216 | |||
NM_138723.1 | NP_620049.1 | 2 | 222 | |||
XM_006719151.3 | XP_006719214.1 | 4 | 446 | |||
XM_011520848.1 | XP_011519150.1 | 4 | 666 | |||
XM_011520849.1 | XP_011519151.1 | 3 | 371 | |||
XM_017019963.1 | XP_016875452.1 | 3 | 191 | |||
GenBank mRNA | AF281254.1 | - | 2 | 216 | ||
AF281255.1 | - | 2 | 216 | |||
AK223415.1 | - | 2 | 207 | |||
AK292995.1 | - | 2 | 227 | |||
AY040274.1 | - | 1 | 28 | |||
BC025778.1 | - | 2 | 222 | 5222875 |