Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NOX5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001184779.1 | NP_001171708.1 | 12 | 1751 | ||
NM_001184780.1 | NP_001171709.1 | 12 | 1878 | |||
NM_024505.3 | NP_078781.3 | 12 | 1835 | |||
NR_033671.2 | 13 | 2066 | ||||
NR_033672.1 | 13 | 1982 | ||||
GenBank mRNA | AF317889.1 | - | 9 | 1266 | ||
AF325189.1 | - | 13 | 1957 | |||
AF325190.1 | - | 13 | 2041 | |||
AF353088.1 | - | 12 | 1730 | |||
AF353089.1 | - | 12 | 1814 | |||
AK026011.1 | - | 9 | 1320 | |||
AK074058.1 | - | 10 | 1504 | |||
AK074071.1 | - | 11 | 4953 | |||
AK314689.1 | - | 12 | 1814 | |||
AY166601.1 | - | 8 | 1061 | |||
BC125097.1 | - | 12 | 1744 | 40118286 | ||
BC125098.1 | - | 13 | 1848 | 40118288 | ||
DQ413001.1 | - | 9 | 1194 |