Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WDR25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001161476.1 | NP_001154948.1 | 3 | 1020 | ||
NM_024515.4 | NP_078791.3 | 3 | 1151 | |||
XM_005268056.4 | XP_005268113.1 | 3 | 1027 | |||
XM_005268057.3 | XP_005268114.1 | 2 | 177 | |||
XM_005268058.4 | XP_005268115.1 | 2 | 191 | |||
XM_011537150.2 | XP_011535452.1 | 3 | 1274 | |||
XM_011537151.2 | XP_011535453.2 | 2 | 630 | |||
XM_017021639.1 | XP_016877128.1 | 2 | 179 | |||
GenBank mRNA | AK291960.1 | - | 3 | 983 | ||
AY295082.1 | - | 3 | 1034 | |||
BC003641.2 | - | 2 | 257 | 3529568 | ||
CT003969.1 | - | 2 | 183 |