Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM65A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001193522.1 | NP_001180451.1 | 22 | 3779 | ||
NM_001193523.1 | NP_001180452.1 | 22 | 3892 | |||
NM_001193524.1 | NP_001180453.1 | 22 | 3805 | |||
NM_024519.3 | NP_078795.2 | 22 | 3770 | |||
XM_011523321.1 | XP_011521623.1 | 22 | 4584 | |||
XM_011523322.1 | XP_011521624.1 | 22 | 3779 | |||
XM_011523324.2 | XP_011521626.1 | 22 | 3839 | |||
XM_011523325.1 | XP_011521627.1 | 22 | 3794 | |||
XM_011523326.1 | XP_011521628.1 | 22 | 3993 | |||
XM_017023663.1 | XP_016879152.1 | 22 | 3776 | |||
XM_017023664.1 | XP_016879153.1 | 21 | 3665 | |||
GenBank mRNA | AB067517.1 | - | 21 | 3625 | ||
AK023787.1 | - | 17 | 3262 | |||
AK127792.1 | - | 22 | 3779 | |||
AK293748.1 | - | 22 | 3802 | |||
AK295677.1 | - | 22 | 3736 | |||
AK302868.1 | - | 11 | 3034 | |||
AL834312.1 | - | 12 | 2835 | |||
BC001850.4 | - | 8 | 1045 | |||
BC054512.1 | - | 12 | 2812 | |||
BC098587.1 | - | 16 | 3207 |