Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C2orf47 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024520.2 | NP_078796.2 | 4 | 878 | ||
XM_005246852.2 | XP_005246909.1 | 3 | 789 | |||
GenBank mRNA | AK026208.1 | - | 4 | 878 | ||
AL832964.1 | - | 3 | 197 | |||
BC017959.1 | - | 3 | 562 | 4607945 |