Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NKAIN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024522.2 | NP_078798.2 | 3 | 565 | ||
GenBank mRNA | AI884789.1 | - | 3 | 281 | ||
AK022712.1 | - | 2 | 137 | |||
AK299507.1 | - | 3 | 237 | |||
BC096707.1 | - | 2 | 123 | 40022279 | ||
BC098173.1 | - | 2 | 123 | 40022282 | ||
BC098249.1 | - | 2 | 123 | 40022276 | ||
BC131518.1 | - | 2 | 123 | 40022275 |