Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RNF219 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024546.3 | NP_078822.3 | 5 | 645 | ||
XM_011535225.1 | XP_011533527.1 | 4 | 495 | |||
XM_017020743.1 | XP_016876232.1 | 5 | 2440 | |||
GenBank mRNA | AK098640.1 | - | 5 | 657 | ||
AL136651.1 | - | 5 | 617 | |||
BC136764.1 | - | 5 | 640 | 9020754 | ||
BC136765.1 | - | 5 | 640 | 9020755 | ||
BI550796.1 | - | 5 | 563 | |||
CR936752.1 | - | 5 | 622 | |||
CR936754.1 | - | 5 | 632 | |||
CR936764.1 | - | 5 | 619 |