Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CEP97 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001303401.1 | NP_001290330.1 | 5 | 599 | ||
NM_024548.3 | NP_078824.2 | 5 | 599 | |||
XM_006713743.3 | XP_006713806.1 | 4 | 990 | |||
XM_011513127.1 | XP_011511429.1 | 5 | 556 | |||
XM_017007178.1 | XP_016862667.1 | 4 | 992 | |||
GenBank mRNA | AK093100.1 | - | 2 | 140 | ||
AK294784.1 | - | 5 | 537 | |||
AK309806.1 | - | 4 | 498 | |||
AL833269.1 | - | 5 | 549 | |||
BC041085.1 | - | 5 | 542 | 5528397 |