Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VCPKMT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040662.1 | NP_001035752.1 | 1, 2 | 304 | ||
NM_024558.2 | NP_078834.2 | 1, 2 | 304 | |||
NR_049738.1 | 1, 2 | 304 | ||||
NR_049739.1 | 1 | 411 | ||||
XM_005268060.3 | XP_005268117.1 | 1, 2 | 290 | |||
XM_017021640.1 | XP_016877129.1 | 1, 2 | 290 | |||
XM_017021641.1 | XP_016877130.1 | 1, 2 | 290 | |||
XM_017021642.1 | XP_016877131.1 | 1 | 387 | |||
GenBank mRNA | AK023982.1 | - | 1, 2 | 304 | ||
AK129564.1 | - | 1 | 290 | |||
BC105118.1 | - | 1, 2 | 325 | 8144121 | ||
BC143673.1 | - | 1, 2 | 325 | 9052187 | ||
BC143674.1 | - | 1, 2 | 325 | 9052188 | ||
BI753530.1 | - | 1, 2 | 284 | |||
BI767189.1 | - | 1, 2 | 275 | |||
BX247997.1 | - | 1 | 390 |