Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C1orf54 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301039.1 | NP_001287968.1 | 6 | 482 | ||
NM_001301040.1 | NP_001287969.1 | 6 | 479 | |||
NM_001301041.1 | NP_001287970.1 | 4 | 268 | |||
NM_024579.3 | NP_078855.2 | 5 | 368 | |||
XM_011509984.2 | XP_011508286.1 | 5 | 398 | |||
GenBank mRNA | AI004135.1 | - | 2 | 172 | ||
AK026874.1 | - | 5 | 353 | |||
AX770997.1 | - | 5 | 366 | |||
AX770998.1 | - | 6 | 396 | |||
BC017761.1 | - | 5 | 353 | 4617936 | ||
DA358536.1 | - | 4 | 279 |