Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM184A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100411.2 | NP_001093881.1 | 16, 17 | 3159 | ||
NM_001288576.1 | NP_001275505.1 | 15, 16 | 3054 | |||
NM_024581.5 | NP_078857.5 | 17, 18 | 3697 | |||
GenBank mRNA | AJ420503.1 | - | 1, 2 | 2177 | ||
AK024004.1 | - | 10, 11 | 1501 | |||
AK091682.1 | - | 15, 16 | 3050 | |||
AK127744.1 | - | 13, 14 | 1999 | |||
AU160036.1 | - | 1, 2 | 345 | |||
AY131226.1 | - | 13, 14 | 1973 | |||
BC009055.1 | - | 9, 10 | 1289 | |||
BC060769.1 | - | 17, 18 | 3697 | 30337859 | ||
BX538186.1 | - | 16, 17 | 3139 |