Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RHBDF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005498.3 | NP_001005498.2 | 7, 8 | 1134 | ||
NM_024599.5 | NP_078875.4 | 8, 9 | 1296 | |||
XM_005257669.3 | XP_005257726.1 | 8, 9 | 1657 | |||
XM_005257670.1 | XP_005257727.1 | 8, 9 | 1166 | |||
XM_011525249.2 | XP_011523551.1 | 8, 9 | 2613 | |||
XM_011525250.2 | XP_011523552.1 | 9, 10 | 1323 | |||
XM_011525251.2 | XP_011523553.1 | 8, 9 | 1140 | |||
XM_017025079.1 | XP_016880568.1 | 8, 9 | 1570 | |||
GenBank mRNA | AK025994.1 | - | 3, 4 | 320 | ||
AK057882.1 | - | 5, 6 | 990 | |||
AK074034.1 | - | 6, 7 | 1601 | |||
AK225684.1 | - | 3, 4 | 273 | |||
AK292135.1 | - | 8, 9 | 1289 | |||
AK292166.1 | - | 7, 8 | 1089 | |||
AK303434.1 | - | 6, 7 | 950 | |||
AL833680.1 | - | 2, 3 | 189 | |||
AY500247.1 | - | 7, 8 | 985 | |||
BC016034.1 | - | 8, 9 | 1276 | 3910543 | ||
BC035829.1 | - | 3, 4 | 382 |