Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HECTD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024602.5 | NP_078878.3 | 10 | 1580 | ||
XM_011542140.2 | XP_011540442.1 | 8 | 977 | |||
GenBank mRNA | AK056545.1 | - | 8 | 931 | ||
AK056873.1 | - | 2 | 474 | |||
AK096462.1 | - | 10 | 1487 | |||
AK301124.1 | - | 9 | 1168 | |||
BC126352.1 | - | 7 | 857 | 8992068 | ||
BC130470.1 | - | 7 | 857 | 40146501 |