Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HECTD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024602.5 | NP_078878.3 | 7, 8 | 1170 | ||
XM_011542140.2 | XP_011540442.1 | 5, 6 | 567 | |||
GenBank mRNA | AK056545.1 | - | 5, 6 | 521 | ||
AK096462.1 | - | 7, 8 | 1077 | |||
AK301124.1 | - | 6, 7 | 758 | |||
BC126352.1 | - | 4, 5 | 447 | 8992068 | ||
BC130470.1 | - | 4, 5 | 447 | 40146501 | ||
BQ215795.1 | - | 6, 7 | 686 |