Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BEND5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001302082.1 | NP_001289011.1 | 5 | 1203 | ||
NM_024603.3 | NP_078879.2 | 5 | 1121 | |||
XM_011542141.2 | XP_011540443.1 | 5 | 3621 | |||
XM_011542142.2 | XP_011540444.1 | 4 | 1069 | |||
XM_017002331.1 | XP_016857820.1 | 6 | 1154 | |||
XM_017002332.1 | XP_016857821.1 | 6 | 1031 | |||
XM_017002334.1 | XP_016857823.1 | 6 | 1250 | |||
GenBank mRNA | AK021650.1 | - | 5 | 1065 | ||
AK055449.1 | - | 3 | 1754 | |||
BC007932.2 | - | 5 | 1073 | 4121760 | ||
BC011804.2 | - | 5 | 887 | 4336159 | ||
BQ006447.1 | - | 2 | 311 | |||
BU743157.1 | - | 2 | 312 | |||
KU178776.1 | - | 5 | 983 | |||
KU178777.1 | - | 5 | 1065 | |||
KU178778.1 | - | 4 | 849 |