Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HSPBAP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320728.1 | NP_001307657.1 | 3 | 465 | ||
NM_024610.5 | NP_078886.2 | 3 | 457 | |||
XM_005247762.2 | XP_005247819.1 | 3 | 457 | |||
XM_011513130.2 | XP_011511432.1 | 4 | 375 | |||
XM_011513132.2 | XP_011511434.2 | 3 | 216 | |||
XM_017007179.1 | XP_016862668.1 | 2 | 281 | |||
GenBank mRNA | AF400663.1 | - | 3 | 424 | ||
AK096705.1 | - | 3 | 352 | |||
AK289410.1 | - | 3 | 457 | |||
BC011897.2 | - | 3 | 441 | 3949150 | ||
BC063629.1 | - | 3 | 397 | |||
BI912690.1 | - | 3 | 437 | |||
HY020808.1 | - | 3 | 468 |