Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FN3KRP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024619.3 | NP_078895.2 | 3 | 404 | ||
NR_046408.1 | 4 | 532 | ||||
GenBank mRNA | AK022233.1 | - | 3 | 371 | ||
AK293523.1 | - | 3 | 388 | |||
AK301958.1 | - | 4 | 516 | |||
AK315373.1 | - | 3 | 388 | |||
AL136631.1 | - | 3 | 375 | |||
AY360465.1 | - | 3 | 371 | |||
BC007611.1 | - | 3 | 380 | 3351601 | ||
BC014408.1 | - | 3 | 358 | 4652087 | ||
CR533461.1 | - | 3 | 354 | |||
DA019669.1 | - | 3 | 405 |