Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FASTKD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001281476.2 | NP_001268405.1 | 6 | 1450 | ||
NM_001322046.1 | NP_001308975.1 | 6 | 2123 | |||
NM_001322048.1 | NP_001308977.1 | 5 | 1378 | |||
NM_001322049.1 | NP_001308978.1 | 5 | 1381 | |||
NM_024622.5 | NP_078898.3 | 6 | 1450 | |||
NR_104020.2 | 6 | 1450 | ||||
XM_006712750.3 | XP_006712813.1 | 5 | 1376 | |||
XM_006712751.3 | XP_006712814.1 | 2 | 1461 | |||
XM_017004911.1 | XP_016860400.1 | 6 | 2763 | |||
XM_017004912.1 | XP_016860401.1 | 2 | 1461 | |||
GenBank mRNA | AB058703.1 | - | 4 | 632 | ||
AK025554.1 | - | 3 | 562 | |||
AK055892.1 | - | 6 | 1361 | |||
AK074302.1 | - | 6 | 1341 | |||
AK130797.1 | - | 6 | 1373 | |||
AK225293.1 | - | 3 | 562 | |||
AL832058.1 | - | 6 | 1333 | |||
BC032687.2 | - | 5 | 779 | 5581443 |