Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FASTKD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001281476.2 | NP_001268405.1 | 6, 7 | 1487 | ||
NM_001322046.1 | NP_001308975.1 | 6, 7 | 2160 | |||
NM_001322048.1 | NP_001308977.1 | 5, 6 | 1415 | |||
NM_001322049.1 | NP_001308978.1 | 5, 6 | 1418 | |||
NM_024622.5 | NP_078898.3 | 6, 7 | 1487 | |||
NR_104020.2 | 6, 7 | 1487 | ||||
XM_006712750.3 | XP_006712813.1 | 5, 6 | 1413 | |||
XM_006712751.3 | XP_006712814.1 | 2, 3 | 1498 | |||
XM_017004911.1 | XP_016860400.1 | 6, 7 | 2800 | |||
XM_017004912.1 | XP_016860401.1 | 2, 3 | 1498 | |||
GenBank mRNA | AB058703.1 | - | 4, 5 | 669 | ||
AK025554.1 | - | 3, 4 | 599 | |||
AK055892.1 | - | 6, 7 | 1398 | |||
AK074302.1 | - | 6, 7 | 1378 | |||
AK130797.1 | - | 6, 7 | 1410 | |||
AK225293.1 | - | 3, 4 | 599 | |||
AL832058.1 | - | 6, 7 | 1370 | |||
BC032687.2 | - | 5, 6 | 816 | 5581443 |