Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CENPU | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024629.3 | NP_078905.2 | 6 | 466 | ||
NR_104593.1 | 6 | 466 | ||||
XM_005263218.4 | XP_005263275.2 | 6 | 501 | |||
GenBank mRNA | AF469667.1 | - | 6 | 469 | ||
AF516710.1 | - | 7 | 883 | |||
AK027121.1 | - | 4 | 276 | |||
AK292297.1 | - | 6 | 412 | |||
BC067802.1 | - | 6 | 406 | |||
BC131556.1 | - | 6 | 466 | 40108113 | ||
BC141854.1 | - | 6 | 464 | 8860370 | ||
DQ907910.1 | - | 6 | 396 | |||
DQ907911.1 | - | 6 | 396 |