Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GALNT12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024642.4 | NP_078918.3 | 10 | 1717 | ||
XM_006717287.1 | XP_006717350.1 | 10 | 1533 | |||
XM_011519018.2 | XP_011517320.1 | 11 | 2389 | |||
XM_011519020.1 | XP_011517322.1 | 10 | 1565 | |||
XM_017015133.1 | XP_016870622.1 | 11 | 1477 | |||
GenBank mRNA | AB040673.1 | - | 10 | 1717 | ||
AB078146.1 | - | 10 | 1717 | |||
AJ132365.2 | - | 10 | 1717 | |||
AJ505963.1 | - | 4 | 496 | |||
AK024865.1 | - | 9 | 1290 |