Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C11orf80 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001302084.1 | NP_001289013.1 | 7 | 666 | ||
NM_024650.3 | NP_078926.3 | 9 | 955 | |||
GenBank mRNA | AK026184.1 | - | 8 | 756 | ||
AK225435.1 | - | 8 | 756 | |||
AK302025.1 | - | 7 | 618 | |||
BC028240.1 | - | 9 | 958 | 5243907 |