Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NOL9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024654.4 | NP_078930.3 | 5 | 1022 | ||
XM_005263493.3 | XP_005263550.1 | 3 | 354 | |||
XM_011542147.2 | XP_011540449.1 | 1 | 46 | |||
GenBank mRNA | AK026976.1 | - | 1 | 32 | ||
AK289653.1 | - | 5 | 954 | |||
BC009257.2 | - | 2 | 112 | |||
BC063639.1 | - | 2 | 62 | |||
BC073860.1 | - | 1 | 25 | |||
BC094836.1 | - | 4 | 397 | |||
BC105095.1 | - | 5 | 1022 | 8144098 | ||
BC110849.1 | - | 5 | 931 | 6167575 | ||
BC112278.1 | - | 5 | 1022 | 8327746 | ||
BC143646.1 | - | 5 | 1022 |