Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BBS10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024685.3 | NP_078961.3 | 2 | 1658 | ||
GenBank mRNA | AK027213.1 | - | 1 | 42 | ||
AK225585.1 | - | 1 | 42 | |||
AK304178.1 | - | 2 | 1631 | |||
BC013795.1 | - | 1 | 250 | |||
BC026355.1 | - | 2 | 1643 | 4817227 |