Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TTLL7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024686.4 | NP_078962.4 | 5 | 712 | ||
XM_005271208.2 | XP_005271265.1 | 4 | 523 | |||
XM_011542159.2 | XP_011540461.1 | 5 | 902 | |||
XM_017002347.1 | XP_016857836.1 | 6 | 732 | |||
XM_017002348.1 | XP_016857837.1 | 5 | 902 | |||
XM_017002349.1 | XP_016857838.1 | 5 | 704 | |||
GenBank mRNA | AK126792.1 | - | 5 | 609 | ||
AK128604.1 | - | 1 | 278 | |||
AY170843.1 | - | 4 | 486 | |||
BC032829.1 | - | 5 | 671 | |||
BC048970.1 | - | 5 | 654 | 5295193 | ||
BC060878.1 | - | 5 | 694 | 5270163 |