Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF419 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098491.1 | NP_001091961.1 | 5 | 580 | ||
NM_001098492.1 | NP_001091962.1 | 4 | 541 | |||
NM_001098493.1 | NP_001091963.1 | 4 | 538 | |||
NM_001098494.1 | NP_001091964.1 | 4 | 481 | |||
NM_001098495.1 | NP_001091965.1 | 3 | 442 | |||
NM_001098496.1 | NP_001091966.1 | 3 | 439 | |||
NM_001291743.1 | NP_001278672.1 | 4 | 478 | |||
NM_001291744.1 | NP_001278673.1 | 4 | 497 | |||
NM_024691.3 | NP_078967.3 | 5 | 577 | |||
GenBank mRNA | AB209371.1 | - | 3 | 3826 | ||
AK026886.1 | - | 4 | 451 | |||
AK302135.1 | - | 3 | 400 | |||
AK304569.1 | - | 4 | 486 | |||
AK307802.1 | - | 5 | 569 | |||
AK309427.1 | - | 3 | 1044 | |||
AK316156.1 | - | 4 | 499 | |||
AK316297.1 | - | 4 | 531 | |||
BC008297.1 | - | 5 | 523 | 3544044 | ||
BC024259.1 | - | 5 | 523 | 3604215 | ||
BP336834.1 | - | 4 | 539 | |||
CT002615.1 | - | 4 | 408 | |||
DA548110.1 | - | 4 | 494 | |||
DC315347.1 | - | 4 | 541 | |||
DC392630.1 | - | 4 | 503 | |||
DC407547.1 | - | 3 | 400 | |||
DC425209.1 | - | 3 | 410 |