Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZFAND1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170796.1 | NP_001164267.1 | 6 | 483 | ||
NM_001170797.1 | NP_001164268.1 | 6 | 483 | |||
NM_024699.2 | NP_078975.2 | 6 | 483 | |||
NR_033193.1 | 5 | 443 | ||||
NR_033194.1 | 5 | 443 | ||||
NR_033195.1 | 5 | 421 | ||||
NR_033196.1 | 5 | 443 | ||||
GenBank mRNA | AK024069.1 | - | 6 | 474 | ||
AK225195.1 | - | 6 | 479 | |||
AL833030.1 | - | 2 | 1011 | |||
BC022251.2 | - | 6 | 468 | 4695118 | ||
BC051857.1 | - | 5 | 425 | 6012307 | ||
BC056877.1 | - | 5 | 426 | 6140929 | ||
BG618233.1 | - | 5 | 426 | |||
BQ427843.1 | - | 6 | 483 | |||
BQ435400.1 | - | 5 | 422 | |||
BU624414.1 | - | 2 | 245 | |||
CA412820.1 | - | 2 | 245 | |||
CR457323.1 | - | 6 | 460 |