Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KATNBL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024713.2 | NP_078989.1 | 5 | 685 | ||
XM_005254668.3 | XP_005254725.1 | 5 | 735 | |||
XM_017022571.1 | XP_016878060.1 | 6 | 769 | |||
XM_017022572.1 | XP_016878061.1 | 6 | 1074 | |||
XM_017022573.1 | XP_016878062.1 | 5 | 895 | |||
XM_017022574.1 | XP_016878063.1 | 6 | 748 | |||
XM_017022575.1 | XP_016878064.1 | 5 | 688 | |||
GenBank mRNA | AK026210.1 | - | 4 | 504 | ||
AK293021.1 | - | 5 | 622 | |||
AK293695.1 | - | 4 | 554 | |||
AK297904.1 | - | 5 | 650 | |||
AK311338.1 | - | 4 | 405 | |||
AL136908.1 | - | 5 | 611 | |||
AL833631.1 | - | 5 | 669 | |||
BC026234.1 | - | 5 | 577 | 4693115 | ||
BC111000.1 | - | 5 | 646 | 5751934 |