Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MCTP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001002796.3 | NP_001002796.1 | 5 | 808 | ||
NM_001297777.1 | NP_001284706.1 | 5 | 808 | |||
NM_024717.5 | NP_078993.4 | 5 | 1087 | |||
XM_005272082.4 | XP_005272139.1 | 6 | 1214 | |||
XM_005272090.4 | XP_005272147.1 | 5 | 587 | |||
XM_006714692.3 | XP_006714755.1 | 6 | 1214 | |||
XM_011543649.2 | XP_011541951.1 | 6 | 1288 | |||
XM_011543650.2 | XP_011541952.1 | 4 | 265 | |||
XM_017009855.1 | XP_016865344.1 | 5 | 527 | |||
XM_017009856.1 | XP_016865345.1 | 5 | 587 | |||
XM_017009857.1 | XP_016865346.1 | 5 | 412 | |||
XM_017009858.1 | XP_016865347.1 | 5 | 527 | |||
XM_017009859.1 | XP_016865348.1 | 5 | 412 | |||
XM_017009860.1 | XP_016865349.1 | 5 | 587 | |||
XM_017009861.1 | XP_016865350.1 | 5 | 922 | |||
XM_017009862.1 | XP_016865351.1 | 5 | 959 | |||
XM_017009863.1 | XP_016865352.1 | 5 | 587 | |||
XM_017009864.1 | XP_016865353.1 | 6 | 1288 | |||
XM_017009865.1 | XP_016865354.1 | 5 | 868 | |||
GenBank mRNA | AK057694.1 | - | 5 | 587 | ||
AK058012.1 | - | 4 | 354 | |||
AK297325.1 | - | 5 | 512 | |||
AY656715.1 | - | 5 | 1087 | |||
AY656716.1 | - | 5 | 424 | |||
BC030005.2 | - | 5 | 569 | 3834212 |