Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GRTP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286732.1 | NP_001273661.1 | 7 | 1000 | ||
NM_024719.3 | NP_078995.2 | 7 | 1000 | |||
XM_005268333.3 | XP_005268390.1 | 6 | 837 | |||
XM_017020755.1 | XP_016876244.1 | 7 | 958 | |||
XM_017020756.1 | XP_016876245.1 | 8 | 1138 | |||
GenBank mRNA | AB449887.1 | - | 7 | 903 | ||
AK026127.1 | - | 5 | 643 | |||
AK300897.1 | - | 7 | 950 | |||
AW243250.1 | - | 2 | 431 | |||
BC033071.2 | - | 5 | 724 | |||
BC062461.1 | - | 6 | 848 | |||
BC080640.1 | - | 7 | 957 | |||
BC112123.1 | - | 5 | 677 | 8327591 | ||
BC112125.1 | - | 5 | 689 | 8327593 | ||
BC143682.1 | - | 4 | 749 | |||
CB242314.1 | - | 2 | 420 |