Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRRC31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001277127.1 | NP_001264056.1 | 9 | 1614 | ||
NM_001277128.1 | NP_001264057.1 | 8 | 1428 | |||
NM_024727.3 | NP_079003.2 | 9 | 1596 | |||
XM_011513158.2 | XP_011511460.1 | 8 | 1428 | |||
XM_011513159.2 | XP_011511461.1 | 8 | 1428 | |||
XM_017007204.1 | XP_016862693.1 | 9 | 1317 | |||
GenBank mRNA | AK026912.1 | - | 8 | 1424 | ||
AY358138.1 | - | 9 | 1526 | |||
BC117401.1 | - | 9 | 1494 | 40125952 | ||
BC143569.1 | - | 9 | 1495 | 9052080 | ||
BC143582.1 | - | 9 | 1512 | |||
EU668335.1 | - | 9 | 1545 |