Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRRC31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001277127.1 | NP_001264056.1 | 5 | 851 | ||
NM_001277128.1 | NP_001264057.1 | 4 | 683 | |||
NM_024727.3 | NP_079003.2 | 5 | 851 | |||
XM_011513158.2 | XP_011511460.1 | 5 | 851 | |||
XM_011513159.2 | XP_011511461.1 | 5 | 851 | |||
XM_011513160.2 | XP_011511462.1 | 5 | 851 | |||
XM_017007204.1 | XP_016862693.1 | 5 | 572 | |||
GenBank mRNA | AK026912.1 | - | 4 | 679 | ||
AK097338.1 | - | 5 | 800 | |||
AY358138.1 | - | 5 | 781 | |||
BC117401.1 | - | 5 | 749 | 40125952 | ||
BC143569.1 | - | 5 | 750 | 9052080 | ||
BC143582.1 | - | 5 | 749 | |||
EU668335.1 | - | 5 | 800 |