Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MYH14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001077186.1 | NP_001070654.1 | 11 | 1199 | ||
NM_001145809.1 | NP_001139281.1 | 11 | 1199 | |||
NM_024729.3 | NP_079005.3 | 10 | 1175 | |||
XM_006723386.3 | XP_006723449.1 | 11 | 1260 | |||
XM_011527320.2 | XP_011525622.1 | 11 | 1277 | |||
XM_011527321.2 | XP_011525623.1 | 10 | 1253 | |||
XM_011527323.2 | XP_011525625.1 | 11 | 1267 | |||
GenBank mRNA | AB290169.1 | - | 10 | 1169 | ||
AK125082.1 | - | 11 | 1278 | |||
AK307783.1 | - | 11 | 1182 | |||
AY165122.1 | - | 10 | 1175 | |||
BC129803.1 | - | 11 | 1053 |