Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KLHL36 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024731.3 | NP_079007.2 | 4 | 1329 | ||
XM_005256149.2 | XP_005256206.1 | 4 | 1543 | |||
XM_017023674.1 | XP_016879163.1 | 4 | 1291 | |||
XM_017023675.1 | XP_016879164.1 | 4 | 1311 | |||
GenBank mRNA | AK022605.1 | - | 3 | 1182 | ||
AK074056.1 | - | 1 | 258 | |||
AK122745.1 | - | 2 | 1764 | |||
BC033821.1 | - | 4 | 1315 | 5219021 |