Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UGT2A3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024743.3 | NP_079019.3 | 5 | 1199 | ||
NR_024010.1 | 6 | 1296 | ||||
XM_011532247.2 | XP_011530549.1 | 5 | 1226 | |||
GenBank mRNA | AK025587.1 | - | 5 | 1004 | ||
AK225308.1 | - | 5 | 1004 | |||
AK293381.1 | - | 6 | 867 | |||
AY358727.1 | - | 5 | 1194 | |||
AY542891.1 | - | 5 | 1199 | |||
BC130533.1 | - | 5 | 1192 | 40146564 |