Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TTC21B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024753.4 | NP_079029.3 | 17 | 2365 | ||
XM_006712761.1 | XP_006712824.1 | 17 | 2308 | |||
XM_011511871.2 | XP_011510173.1 | 12 | 5610 | |||
XM_011511872.2 | XP_011510174.1 | 17 | 2308 | |||
XM_017004967.1 | XP_016860456.1 | 17 | 2308 | |||
XM_017004968.1 | XP_016860457.1 | 12 | 3856 | |||
XM_017004969.1 | XP_016860458.1 | 15 | 2025 | |||
GenBank mRNA | BC055424.1 | - | 17 | 2365 | 4837489 | |
BX647903.1 | - | 3 | 116 |