Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CAMKMT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024766.4 | NP_079042.1 | 10 | 902 | ||
XM_011533113.2 | XP_011531415.1 | 8 | 578 | |||
XM_017004982.1 | XP_016860471.1 | 8 | 3005 | |||
XM_017004983.1 | XP_016860472.1 | 9 | 1117 | |||
GenBank mRNA | AK027104.1 | - | 10 | 842 | ||
BC053733.1 | - | 10 | 829 | 5199179 |