Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NAA40 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001300800.1 | NP_001287729.1 | 6 | 669 | ||
NM_024771.3 | NP_079047.2 | 6 | 598 | |||
XM_006718689.3 | XP_006718752.1 | 6 | 495 | |||
XM_011545254.2 | XP_011543556.1 | 7 | 672 | |||
XM_011545255.2 | XP_011543557.1 | 7 | 582 | |||
GenBank mRNA | AK023910.1 | - | 6 | 567 | ||
AK299041.1 | - | 6 | 663 | |||
AK300451.1 | - | 5 | 519 | |||
AK303557.1 | - | 4 | 429 | |||
BC041617.1 | - | 6 | 527 | 5921480 | ||
BC052298.1 | - | 6 | 527 | 6292760 | ||
BX647309.1 | - | 5 | 1624 | |||
JF263566.1 | - | 6 | 567 |