Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: QSER1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001076786.2 | NP_001070254.1 | 4 | 2556 | ||
XM_006718325.3 | XP_006718388.3 | 4 | 2908 | |||
XM_017018327.1 | XP_016873816.1 | 4 | 2908 | |||
XM_017018328.1 | XP_016873817.1 | 4 | 2908 | |||
XM_017018329.1 | XP_016873818.1 | 2 | 3361 | |||
XM_017018330.1 | XP_016873819.1 | 3 | 2306 | |||
GenBank mRNA | AK123858.1 | - | 1 | 618 | ||
AK125391.1 | - | 1 | 1978 | |||
AK125902.1 | - | 1 | 104 | |||
AK126023.1 | - | 5 | 1860 | |||
AL834141.1 | - | 1 | 1251 | |||
BC112935.1 | - | 4 | 2556 |