Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LONRF3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001031855.2 | NP_001027026.1 | 2 | 1078 | ||
NM_001289109.1 | NP_001276038.1 | 2 | 708 | |||
NM_024778.5 | NP_079054.3 | 2 | 1078 | |||
NR_110311.1 | 2 | 1078 | ||||
XM_005262476.2 | XP_005262533.1 | 2 | 1078 | |||
XM_011531398.2 | XP_011529700.1 | 2 | 1078 | |||
XM_017029845.1 | XP_016885334.1 | 2 | 1078 | |||
GenBank mRNA | AK026265.1 | - | 2 | 330 | ||
AK091777.1 | - | 2 | 1074 | |||
AK097641.1 | - | 2 | 708 | |||
AK290198.1 | - | 2 | 1074 | |||
BC099847.1 | - | 2 | 942 | 40007729 | ||
BC100671.1 | - | 2 | 942 | 40007726 | ||
BC103491.1 | - | 2 | 942 | 40007723 |